MilliporeSigma
  • Fabrication of novel magnetic chitosan grafted with graphene oxide to enhance adsorption properties for methyl blue.
Fabrication of novel magnetic chitosan grafted with graphene oxide to enhance adsorption properties for methyl blue.
Journal of hazardous materials (2012-03-21)
Lulu Fan, Chuannan Luo, Xiangjun Li, Fuguang Lu, Huamin Qiu, Min Sun
ABSTRACT
A novel magnetic composite bioadsorbent composed of magnetic chitosan and graphene oxide (MCGO) was prepared as the magnetic adsorbent. The morphology, chemical structure and magnetic property of the MCGO were characterized by Fourier transform infrared spectrometer (FT-IR), X-ray diffraction (XRD) and Scanning electronic microscope (SEM), respectively. Adsorption of methyl blue (MB) onto MCGO was investigated with respect to pH, adsorption time, initial MB concentration and temperature. Kinetics data and adsorption isotherm, obtained at the optimum pH 5.3, were better fitted by pseudo-second-order kinetic model and by Langmuir isotherm, respectively. The values of activation parameters such as free energy (ΔG, -0.74∼-1.46kJmol(-1)), enthalpy (ΔH, -10.28kJmol(-1)) and entropy (ΔS, -36.35Jmol(-1)K(-1)) were determined, respectively, indicating that the adsorption was spontaneous, favorable and exothermic process in nature. Moreover, the MCGO was stable and easily recovered, the adsorption capacity was about 90% of the initial saturation adsorption capacity after being used four times.
Sigma-Aldrich
Methyl Blue, powder or crystals
Sigma-Aldrich
Methyl Blue, certified by the BSC
Sigma-Aldrich
Methyl Blue, for microscopy (Bot., Hist.), indicator (pH 9.4-14.0)