MilliporeSigma
  • Localization of Islet-1-positive cells in the healthy and infarcted adult murine heart.
Localization of Islet-1-positive cells in the healthy and infarcted adult murine heart.
Circulation research (2012-03-20)
Florian Weinberger, Dennis Mehrkens, Felix W Friedrich, Mandy Stubbendorff, Xiaoqin Hua, Jana Christina Müller, Sonja Schrepfer, Sylvia M Evans, Lucie Carrier, Thomas Eschenhagen
ABSTRACT
The transcription factor Islet-1 is a marker of cardiovascular progenitors during embryogenesis. The isolation of Islet-1-positive (Islet-1(+)) cells from early postnatal hearts suggested that Islet-1 also marks cardiac progenitors in adult life. We investigated the distribution and identity of Islet-1(+) cells in adult murine heart and evaluated whether their number or distribution change with age or after myocardial infarction. Distribution of Islet-1(+) cells in adult heart was investigated using gene targeted mice with nuclear β-galactosidase inserted into the Islet-1 locus. nLacZ-positive cells were only present in 3 regions of the adult heart: clusters in the interatrial septum and around the pulmonary veins, scattered within the wall of the great vessels, and a strictly delimited cluster between the right atrium and superior vena cava. Islet-1(+) cells in the first type of clusters coexpressed markers for parasympathetic neurons. Positive cells in the great arteries coexpressed smooth muscle actin and myosin heavy chain, indicating a smooth muscle cell identity. Very few Islet-1(+) cells within the outflow tract expressed the cardiomyocyte marker α-actinin. Islet-1(+) cells in the right atrium coexpressed the sinoatrial node pacemaker cell marker HCN4. Cell number and localization remained unchanged between 1 to 18 months of age. Consistently Islet-1 mRNA was detected in human sinoatrial node. Islet-1(+) cells could not be detected in the infarct zone 2 to 28 days after myocardial infarction, aside from 10 questionable cells in 1/13 hearts. Our results identify Islet-1 as a novel marker of the adult sinoatrial node and do not provide evidence for Islet-1(+) cells to serve as cardiac progenitors.
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, tablet
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, ≥98%
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, ≥98%, powder
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat