MilliporeSigma
  • Myoglobin-directed assemblies of binary monolayers functionalized with iminodiacetic acid ligands at the air-water interface through metal coordination for multivalent protein binding.
Myoglobin-directed assemblies of binary monolayers functionalized with iminodiacetic acid ligands at the air-water interface through metal coordination for multivalent protein binding.
Physical chemistry chemical physics : PCCP (2012-03-15)
Xiaoyu Wang, Xuan Huang, Yanyan Xin, Xuezhong Du
ABSTRACT
Myoglobin binding to the binary monolayers composed of sodium hexadecylimino diacetate and hexadecanol at the air-water interface by means of metal coordination has been investigated using infrared reflection absorption spectroscopy (IRRAS). In the absence of Cu(2+), no myoglobin binding to the binary monolayers was observed. In the presence of Cu(2+), remarkable myoglobin binding to the binary monolayers resulted from the formation of ternary complexes of iminodiacetate (IDA)-Cu(2+)-surface histidine. Myoglobin-directed assemblies of the binary monolayers facilitated multivalent protein binding through lateral rearrangements of the IDA ligands and reorientations of the alkyl chains for enhanced protein binding. Myoglobin binding to and desorption from the binary monolayers could be readily controlled through metal coordination.
Sigma-Aldrich
1-Hexadecanol, 95%
Sigma-Aldrich
1-Hexadecanol, ≥99%
Sigma-Aldrich
1-Hexadecanol, ReagentPlus®, 99%
Supelco
Cetyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cetyl alcohol, analytical standard
Sigma-Aldrich
Iminodiacetic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Iminodiacetic acid, 98%
Cetyl alcohol, European Pharmacopoeia (EP) Reference Standard