MilliporeSigma
  • Pretreatment with AT-101 enhances tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)-induced apoptosis of breast cancer cells by inducing death receptors 4 and 5 protein levels.
Pretreatment with AT-101 enhances tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)-induced apoptosis of breast cancer cells by inducing death receptors 4 and 5 protein levels.
Journal of cancer research and clinical oncology (2012-03-14)
Asli Kisim, Harika Atmaca, Burcu Cakar, Bulent Karabulut, Canfeza Sezgin, Selim Uzunoglu, Ruchan Uslu, Burcak Karaca
ABSTRACT
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a member of the TNF superfamily and has been shown to induce extrinsic pathway of apoptosis in many types of cancer cells. AT-101, an (-)-enantiomer of gossypol, is a potent anticancer agent that is shown to be an inhibitor of Bcl-2/Bcl-XL. In this study, we searched whether pretreatment with either of these drugs would result in the enhancement of apoptosis through induction of death receptors and activation of mitochondrial pathways within breast cancer cells. Human breast cancer (MCF-7 and MDA-MB-231) and normal breast cells (MCF-10A) were treated with drugs alone/in combination/sequentially. XTT cell viability assay was used to evaluate cytotoxicity. For showing apoptosis, both DNA Fragmentation and caspase 3/7 activity measurements were done. ELISA and Western blot analysis were done to assess DR4 and DR5 protein levels. The expression levels of apoptotic proteins were assessed by human apoptosis antibody array. The sequential treatment of AT-101 followed by TRAIL resulted in significant synergistic cytotoxicity and apoptosis. Moreover, pretreatment of breast cancer cells with AT-101 and then with TRAIL caused enhancement of the expression levels of DR4 and DR5 in both cancer cell lines, suggesting that these cells were under strong apoptotic stimuli. These findings all together, strongly suggest that pretreatment with AT-101 enhances TRAIL-induced death-inducing signaling complex resulting in the engagement of the mitochondrial pathway to apoptosis in breast cancer cells. These promising, preliminary results make AT-101 and TRAIL a novel combination treatment candidate for breast cancer.
Product Description
Supelco
(±)-Gossypol-acetic acid, analytical standard, crystalline complex consisting of equimolar quantities of gossypol and acetic acid, ~95%