MilliporeSigma
  • Isolation and characterization of multidrug-resistant Klebsiella spp. isolated from shrimp imported from Thailand.
Isolation and characterization of multidrug-resistant Klebsiella spp. isolated from shrimp imported from Thailand.
International journal of food microbiology (2012-03-13)
Mohamed Nawaz, S A Khan, Q Tran, K Sung, A A Khan, I Adamu, R S Steele
ABSTRACT
A study was undertaken to isolate and characterize tetracycline and nalidixic acid-resistant Klebsiella spp. in farm-raised, imported shrimp sold in the United States. Sixty-seven multiple antibiotic-resistant Klebsiella spp. strains were isolated from imported shrimp samples. Using morphological and biochemical methods, fifty-three strains were tentatively identified as Klebsiella pneumoniae and fourteen as K. oxytoca. Although all isolates were resistant to tetracycline, only 8 were resistant to nalidixic acid. These 8 isolates were further screened by PCR for quinolone resistance genes (qnrA, B, S, gyrA, B and parC). PCR protocols failed to amplify any qnr genes. The purified PCR amplicons of gyrA, gyrB and parC were sequenced and analyzed for point mutations that confer resistance to fluoroquinolone antibiotics. Analysis of the sequences of the gyrA amplicons from nalidixic acid-resistant Klebsiella spp. indicated two point mutations in gyrA at positions 83 (Ser→Phe) and 87 (Asp→Ala). Sequence analysis of the parC amplicons indicated an amino acid change at position 80 (Ser→Ile). No mutations were detected in gyrB. Template DNA from all isolates was screened for tetracycline resistance genes (tetA-E). Oligonucleotide primers specifically targeting a 305-bp region of tetB and a 477-bp region of tetD successfully amplified sequences from 91.0 and 44.0% of the isolates, respectively. None of the isolates contained tetA, tetC or tetE genes. Plasmids (2.0-16.0kb) were found in 23 of the 67 isolates. XbaI-PFGE identified 32 distinct macro restriction patterns (mrps) among the 61 multiple drug-resistant Klebsiella spp. that were typable. Our results indicate that imported shrimp is a reservoir for multidrug resistant Klebsiella spp. and potential health risks posed by such strains should not be underestimated.
Sigma-Aldrich
Nalidixic acid sodium salt, powder
Sigma-Aldrich
Nalidixic acid, ≥98%