MilliporeSigma
  • WO3–x@Au@MnO2 core–shell nanowires on carbon fabric for high-performance flexible supercapacitors.
WO3–x@Au@MnO2 core–shell nanowires on carbon fabric for high-performance flexible supercapacitors.
Advanced materials (Deerfield Beach, Fla.) (2012-03-10)
Xihong Lu, Teng Zhai, Xianghui Zhang, Yongqi Shen, Longyan Yuan, Bin Hu, Li Gong, Jian Chen, Yihua Gao, Jun Zhou, Yexiang Tong, Zhong Lin Wang
ABSTRACT
WO3–x@Au@MnO2 core–shell nanowires (NWs) are synthesized on a flexible carbon fabric and show outstanding electrochemical performance in supercapacitors such as high specific capacitance, good cyclic stability, high energy density, and high power density. These results suggest that the WO3–x@Au@MnO2 NWs have promising potential for use in high-performance flexible supercapacitors.
Sigma-Aldrich
Manganese(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Manganese(IV) oxide, activated, ~85%, <10 μm
Sigma-Aldrich
Manganese(IV) oxide, 10 μm, reagent grade, ≥90%
Sigma-Aldrich
Manganese(IV) oxide, ReagentPlus®, ≥99%