MilliporeSigma
  • Open-label evaluation of the skin-brightening efficacy of a skin-brightening system using decapeptide-12.
Open-label evaluation of the skin-brightening efficacy of a skin-brightening system using decapeptide-12.
Journal of cosmetic and laser therapy : official publication of the European Society for Laser Dermatology (2012-03-10)
Andrea T Kassim, Mussarrat Hussain, David J Goldberg
ABSTRACT
This prospective study evaluated the safety and efficacy of decapeptide-12 in conjunction with an antioxidant cleanser, glycolic-acid containing facial moisturizer and broad-spectrum sunscreen in the treatment of facial hyperpigmentation associated with chronic photodamage. Fifteen female subjects with Fitzpatrick skin types I through IV and documented photodamage were entered into the study, of whom 13 completed the study. Results were obtained at weeks 4, 8, 12, 18 and 24 and were assessed by both volunteers and investigators based on standardised digital photography using the Global Assessment of Photodamage Severity Scale. At the conclusion of the study at 24 weeks, 38.5% of the volunteers achieved complete clearance from a moderate (grade 3) degree of photodamage at baseline to completely cleared (grade 1). Another 30.7% improved from a moderate (grade 3) degree of photodamage at baseline to a mild (grade 2) degree of photodamage. An additional 15.4% improved from a severe (grade 4) degree of photodamage at baseline to a moderate (grade 3) degree of photodamage while an equal 15.4% improved from a severe (grade 4) degree of photodamage at baseline to a designation of mild (grade 2) at 24 weeks. All study-related treatments were well tolerated. The mechanism of action of decapeptide-12 includes inhibition of melanin synthesis via inhibition of the tyrosinase enzyme. Advantages of decapeptide-12 over other skin-brightening agents include its low incidence of side effects, lack of cytotoxicity and safe use in ethnic skin as well as in patients who have failed other treatment regimens.
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland