MilliporeSigma
  • Combination of essential oil of Melaleuca alternifolia and iodine in the treatment of molluscum contagiosum in children.
Combination of essential oil of Melaleuca alternifolia and iodine in the treatment of molluscum contagiosum in children.
Journal of drugs in dermatology : JDD (2012-03-08)
Eric Markum, John Baillie
ABSTRACT
Molluscum contagiosum is a common childhood viral skin condition and is increasingly found as a sexually transmitted disease in adults. Current treatment options are invasive, requiring tissue destruction and attendant discomfort. Fifty-three children (mean age 6.3+5.1 years) with the diagnosis of molluscum contagiosum were treated with twice daily topical application of either essential oil of Melaleuca alternifolia (TTO), a combination of TTO and organically bound iodine (TTO-I), or iodine alone. At the end of 30 days, 48 children were available for follow up. A greater than 90% reduction in the number of lesions was observed in 16 of 19 children treated with TTO-I, while 1 of 16 and 3 of 18 children met the same criteria for improvement in the iodine and TTO groups (P<0.01, ANOVA) respectively by intention-to-treat analysis. No child discontinued treatment due to adverse events. The combination of essential oil of M. alternifolia with organically bound iodine offers a safe therapeutic alternative in the treatment of childhood molluscum. Clinical Trial Registry ACTRN12610000984099.
Sigma-Aldrich
Tea tree oil, FG