MilliporeSigma
  • Nicotine, cotinine, and myosmine determination using polymer films of tailor-designed zinc porphyrins as recognition units for piezoelectric microgravimetry chemosensors.
Nicotine, cotinine, and myosmine determination using polymer films of tailor-designed zinc porphyrins as recognition units for piezoelectric microgravimetry chemosensors.
Analytical chemistry (2012-03-08)
Krzysztof Noworyta, Wlodzimierz Kutner, Channa A Wijesinghe, Serge G Srour, Francis D'Souza
ABSTRACT
Two electropolymerizable zinc porphyrins with receptor sites tailor-designed for selective recognition of the nicotine, cotinine, or myosmine alkaloids were synthesized. These were 5-(2-phenoxyacetamide)-10,15,20-tris(triphenylamino)porphyrinato zinc(II) 1 and 5-(2,5-phenylene-bis(oxy)diacetamide)-10,15,20-tris(triphenylamino)porphyrinato zinc(II) 2 featuring one and two pendant amide side "pincers", respectively, and three triphenylamine substituents at the meso positions of the porphyrin macrocycles capable of electrochemical polymerization. Thin polymerfilms of these porphyrins served for recognition and the piezoelectric microgravimetry (PM) for analytical signal transduction of a new chemical sensor devised for determination of these alkaloids. The films were deposited by potentiodynamic electropolymerization on the 10 MHz quartz resonators of the electrochemical quartz crystal microbalance (EQCM) without affecting the electronic structure of the porphyrin macrocycles. Under favorable flow injection analysis (FIA) conditions, the alkaloid analytes were determined at the concentration level of 0.1 mM with high sensitivity and selectivity. Affinity toward the analytes of the polymer of 2 was higher than that of 1 due to the higher binding ability offered by two pendant pincers of the former. Because of the selective receptors and PM applied under FIA conditions, the developed procedure offered an alternative to the time-consuming and relatively expensive high-performance liquid chromatography (HPLC), capillary electrophoresis (CE), and gas chromatography mass spectrometry (GC-MS) methods of detection and quantification of these alkaloids.
Sigma-Aldrich
Myosmine, ≥98%
Pricing and availability is not currently available.