MilliporeSigma
  • Total synthesis of (-)-kaitocephalin based on a Rh-catalyzed C-H amination.
Total synthesis of (-)-kaitocephalin based on a Rh-catalyzed C-H amination.
Organic letters (2012-03-07)
Keisuke Takahashi, Daisuke Yamaguchi, Jun Ishihara, Susumi Hatakeyama
ABSTRACT
A total synthesis of (-)-kaitocephalin, an ionotropic glutamate receptor antagonist, is accomplished in highly stereocontrolled manner via Overman rearrangement, rhodium-catalyzed benzylic C-H amination, pyrrolidine formation involving nucleophilic opening of a cyclic sulfamate, and rhodium-catalyzed allylic C-H amination as key steps.
Product Description
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Sigma-Aldrich
Sulfamic acid, ≥99.5% (alkalimetric)
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%