MilliporeSigma
  • Rapid identification of Branhamella catarrhalis with 4-methylumbelliferyl butyrate.
Rapid identification of Branhamella catarrhalis with 4-methylumbelliferyl butyrate.
Journal of clinical microbiology (1988-06-01)
M Vaneechoutte, G Verschraegen, G Claeys, P Flamen
ABSTRACT
Branhamella catarrhalis can be distinguished from Neisseria spp. by the presence of butyrate esterase. This enzyme can be rapidly detected when 4-methylumbelliferyl butyrate is used as the substrate. All B. catarrhalis strains tested gave a positive fluorescence reaction within 5 min, while Neisseria spp. remained negative, even after 18 h of incubation.
Sigma-Aldrich
4-Methylumbelliferyl butyrate, suitable for fluorescence, ≥95% (HPCE)
Pricing and availability is not currently available.