MilliporeSigma
  • Acute toxicity of butachlor and atrazine to freshwater green alga Scenedesmus obliquus and cladoceran Daphnia carinata.
Acute toxicity of butachlor and atrazine to freshwater green alga Scenedesmus obliquus and cladoceran Daphnia carinata.
Ecotoxicology and environmental safety (2012-03-03)
Hongzhi He, Jing Yu, Guikui Chen, Wenyang Li, Jinbo He, Huashou Li
ABSTRACT
Both single and joint toxicity of atrazine and butachlor to freshwater green alga Scenedesmus obliquus and cladoceran Daphnia carinata isolated from South China were investigated in the present study. The 96 h-EC(50) values of atrazine and butachlor to S. obliquus were 0.0147 and 2.31 mg L(-1), while the 48 h-LC(50) values to D. carinata were 60.6 and 3.40 mg L(-1), respectively. These results suggest that atrazine could be highly toxic to S. obliquus and slightly toxic to D. carinata, while butachlor exhibits moderate toxicity to both organisms. The additive indexes of atrazine and butachlor mixtures were -2.68 (-3.02 to -2.32) to S. obliquus and 0.054 (-0.025 to 0.238) to D. carinata, respectively. Therefore, the joint action of two herbicides was significant antagonism to S. obliquus, while significant synergism was not shown to D. carinata. Moreover, significant linear correlation between the natural logarithm of herbicide concentrations and growth rates of alga S. obliquus was observed. Taken together, it is the first study reporting the toxicity endpoints for mixture of atrazine and butachlor against S. obliquus and D. carinata isolated from south China. The present results would be helpful to provide data to assess the ecological risk of both herbicides to aquatic organisms.
Product Description
Supelco
Butachlor, PESTANAL®, analytical standard