MilliporeSigma
  • New steroidal glycoside ester and aliphatic acid from the fruits of Lycium chinense.
New steroidal glycoside ester and aliphatic acid from the fruits of Lycium chinense.
Journal of Asian natural products research (2012-03-02)
Woo-Suk Jung, Ill-Min Chung, Mohd Ali, Ateeque Ahmad
ABSTRACT
Two new compounds stigmast-5-en-3β-ol-3-O-β-D-(2'-n-triacontanoyl) glucopyranoside (1) and 19,21-dimethyl triacont-17,22,24,26,28-pentaene-1-oic acid (2), along with the three known compounds n-tetracosanyl octadec-9-enoate (3), β-sitosterol, and β-sitosterol-3-O-β-D-glucoside, have been isolated from the methanol extract of Lycium chinense fruits. The structures of these phytoconstituents have been established on the basis of spectral data analysis and chemical reactions.
Sigma-Aldrich
Stigmasterol, ~95%