MilliporeSigma
  • Synthesis and comparative evaluation of 4-oxa- and 4-aza-podophyllotoxins as antiproliferative microtubule destabilizing agents.
Synthesis and comparative evaluation of 4-oxa- and 4-aza-podophyllotoxins as antiproliferative microtubule destabilizing agents.
Bioorganic & medicinal chemistry letters (2012-03-01)
Natalia B Chernysheva, Dmitry V Tsyganov, Alex A Philchenkov, Michael P Zavelevich, Alex S Kiselyov, Roman V Semenov, Marina N Semenova, Victor V Semenov
ABSTRACT
A series of novel 4-oxa-podophyllotoxin derivatives 7 featuring the intact lactone ring D and various substituents in rings B and E has been synthesized and evaluated in a phenotypic sea urchin embryo assay along with the representative 4-aza-analogs 5 for their antimitotic and microtubule destabilizing activity. The most active compounds exhibited myristicin-derived or a 3',5'-dimethoxy substitution pattern in the ring E and a 6-methoxy moiety replacing the methylenedioxy ring A. Compounds 5xb, 5xe, 5yb, 7xa, 7xb, and 7xc showed potent antiproliferative effects in the NCI60 cytotoxicity screen. Notably, growth of the multi-drug resistant NCI/ADR-RES cells was more affected by these agents than the parent OVCAR-8 cell line. Although generally 4-oxa-podophyllotoxins were less potent than the respective 4-aza-derivatives in these assays, stability of the former series towards oxidation may prove to be of interest for the development of anticancer agents with in vivo activity.
Product Description
Supelco
Myristicin, analytical standard
Sigma-Aldrich
Myristicin from parsley leaf oil, ≥85% (HPLC), oil