MilliporeSigma
  • Recoil frame photoelectron angular distributions of BF3: a sensitive probe of the shape resonance in the F 1s continuum.
Recoil frame photoelectron angular distributions of BF3: a sensitive probe of the shape resonance in the F 1s continuum.
The Journal of chemical physics (2012-03-01)
T Mizuno, J Adachi, N Miyauchi, M Kazama, M Stener, P Decleva, A Yagishita
ABSTRACT
Recoil frame photoelectron angular distributions (RFPADs) of BF(3) molecules are presented over the energy region of the shape resonance in the F 1s continuum. Time-dependent density functional theory calculations are also given to understand the shape resonance dynamics. The RFPADs have been compared with the theoretical calculations. It is found that the RFPADs calculated by the localized core-hole model are in better agreement with the experimental, compared with those by the delocalized core hole. Dipole matrix elements and dipole prepared continuum wavefunctions show that the shape resonance in the F 1s ionization continuum of BF(3) is induced by p-partial waves as previously reported by Swanson et al. [J. Chem. Phys. 75, 619 (1981)]. However, due to the couplings with the other partial waves the feature characteristic of the p-partial waves has not been observed in the RFPADs.
Product Description
Supelco
Boron trifluoride - 1-butanol solution, ~10% in 1-butanol (∼1.3 M), for GC derivatization, LiChropur