MilliporeSigma
  • A mutant leucine aminopeptidase from Streptomyces cinnamoneus with enhanced L-aspartyl L-amino acid methyl ester synthetic activity.
A mutant leucine aminopeptidase from Streptomyces cinnamoneus with enhanced L-aspartyl L-amino acid methyl ester synthetic activity.
Biotechnology letters (2012-02-23)
Jiro Arima, Mirai Kono, Manami Kita, Nobuhiro Mori
ABSTRACT
L-aspartyl L-amino acid methyl ester was synthesized using a mutant of a thermostable leucine aminopeptidase from Streptomyces cinnamoneus, D198 K SSAP, obtained in previously. A peptide of high-intensity sweetener, L-aspartyl-L-phenylalanine methyl ester, was selected as a model for demonstrating the synthesis of L-aspartyl L-amino acid methyl ester. The hydrolytic activities of D198 K SSAP toward L-aspartyl-L-phenylalanine and its methyl ester were, respectively, 74-fold and fourfold higher than those of wild type. Similarly, the initial rate of the enzyme for L-aspartyl-L-phenylalanine methyl ester synthesis was over fivefold higher than that of wild-type SSAP in 90% methanol (v/v) in a one-pot reaction. Furthermore, other L-aspartyl L-amino acid methyl esters were synthesized efficiently using D198 K SSAP. Results show that the substitution of Asp198 of SSAP with Lys is effective for synthesizing L-aspartyl L-amino acid methyl ester.
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
Aspartame, European Pharmacopoeia (EP) Reference Standard
Supelco
Aspartame, analytical standard