MilliporeSigma
Antiproliferative effect of α-mangostin on canine osteosarcoma cells.
Research in veterinary science (2012-02-22)
Aungkana Krajarng, Sirinun Nilwarankoon, Sunit Suksamrarn, Ramida Watanapokasin
ABSTRACT
Osteosarcoma is the most frequently diagnosed primary bone tumor in dog. Since chemotherapeutics are quite limited due to high cost and severe toxicity, therefore, the ultimate goal is to discover cost-effective therapeutics with less toxicity. We have studied the effect of α-mangostin, a xanthone derivative isolated from pericarp of mangosteen (Garcinia mangostana Linn.) in canine osteosarcoma, D-17 cells. The results showed that α-mangostin induced antiproliferation with IC(50) at 15 μg/ml. Hoechst 33342 nuclear staining and nucleosomal DNA-gel electrophoresis revealed that α-mangostin could induce nuclear condensation and fragmentation, typically seen in apoptosis. Cell cycle analysis demonstrated that α-mangostin induced sub-G1 peak. In addition, α-mangostin also induced membrane flipping of the phosphatidylserine and the loss of mitochondrial membrane potential in D-17 cells. In conclusion, α-mangostin, induced apoptotic cell death against canine osteosarcoma D-17 cells, could be a potential candidate for preventive and therapeutic application for bone cancer treatment in dogs.
Sigma-Aldrich
α-Mangostin, ≥98% (HPLC)