MilliporeSigma
  • Kinetics of the oxidation of sucralose and related carbohydrates by ferrate(VI).
Kinetics of the oxidation of sucralose and related carbohydrates by ferrate(VI).
Chemosphere (2012-02-22)
Virender K Sharma, Mary Sohn, George A K Anquandah, Nasri Nesnas
ABSTRACT
The kinetics of the oxidation of sucralose, an emerging contaminant, and related monosaccharides and disaccharides by ferrate(VI) (Fe(VI)) were studied as a function of pH (6.5-10.1) at 25°C. Reducing sugars (glucose, fructose, and maltose) reacted faster with Fe(VI) than did the non-reducing sugar sucrose or its chlorinated derivative, sucralose. Second-order rate constants of the reactions of Fe(VI) with sucralose and disaccharides decreased with an increase in pH. The pH dependence was modeled by considering the reactivity of species of Fe(VI), (HFeO(4)(-) and FeO(4)(2-)) with the studied substrates. Second-order rate constants for the reaction of Fe(VI) with monosaccharides displayed an unusual variation with pH and were explained by considering the involvement of hydroxide in catalyzing the ring opening of the cyclic form of the carbohydrate at increased pH. The rate constants for the reactions of carbohydrates with Fe(VI) were compared with those for other oxidant species used in water treatment and were briefly discussed.
Product Description
Supelco
Sucralose, analytical standard
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)