MilliporeSigma
  • Stereoselective access to fluorinated and non-fluorinated quaternary piperidines: synthesis of pipecolic acid and iminosugar derivatives.
Stereoselective access to fluorinated and non-fluorinated quaternary piperidines: synthesis of pipecolic acid and iminosugar derivatives.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-02-16)
Santos Fustero, Laia Albert, Natalia Mateu, Gema Chiva, Javier Miró, Javier González, José Luis Aceña
ABSTRACT
The preparation of optically pure quaternary piperidines, both fluorinated and non-fluorinated, has been achieved from a chiral imino lactone derived from (R)-phenylglycinol. In the case of the fluorinated derivatives, the addition of (trifluoromethyl)trimethylsilane (TMSCF(3)) followed by iodoamination and migration of the CF(3) group allowed access to four derivatives of α-(trifluoromethyl)pipecolic acid. A theoretical study of the CF(3)-group rearrangement has been carried out to help establish the reaction mechanism of this uncommon transformation. Moreover, a route to trifluoromethyl-substituted iminosugars was also developed through the diastereoselective dihydroxylation of suitable synthetic intermediates. Conversely, alkylation of the starting substrate and subsequent cross-metathesis and aza-Michael reactions led to α-alkyl derivatives of the target compounds.
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
L-Pipecolic acid, 99% (titration)