MilliporeSigma
  • Impact of the herbicide (RS)-MCPP on an anaerobic membrane bioreactor performance under different COD/nitrate ratios.
Impact of the herbicide (RS)-MCPP on an anaerobic membrane bioreactor performance under different COD/nitrate ratios.
Bioresource technology (2012-02-10)
Ali Yuzir, Shreeshivadasan Chelliapan, Paul J Sallis
ABSTRACT
The degradation of (RS)-MCPP was investigated in an anaerobic membrane bioreactor (AnMBR) using nitrate as an available electron acceptor under different COD/NO(3)(-)-N ratios. Results showed high soluble COD removal efficiency (80-93%) when the reactor was operated at high COD/NO(3)(-)-N ratios. However, the COD removal started to decline (average 15%) at high nitrate concentrations coinciding with a drop in nitrate removal efficiency to 37%, suggesting that the denitrification activity dropped and affected the AnMBR performance when nitrate was the predominant electron acceptor. Additionally, the removal efficiency of (RS)-MCPP increased from 2% to 47% with reducing COD/NO(3)(-)-N ratios, whilst the (RS)-MCPP specific utilisation rate (SUR) was inversely proportional to the COD/NO(3)(-)-N ratio, suggesting that a lower COD/NO(3)(-)-N ratios had a positive influence on the (RS)-MCPP SUR. Although nitrate had a major impact on methane production rates, the methane composition was stable (approximately 80%) for COD/NO(3)(-)-N ratios of 23 or more.
Product Description
Supelco
MCPA, PESTANAL®, analytical standard
Supelco
Mecoprop-P, PESTANAL®, analytical standard
Sigma-Aldrich
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
Supelco
Mecoprop, PESTANAL®, analytical standard