MilliporeSigma
  • Maternal exposure to drinking-water chlorination by-products and small-for-gestational-age neonates.
Maternal exposure to drinking-water chlorination by-products and small-for-gestational-age neonates.
Epidemiology (Cambridge, Mass.) (2012-02-10)
Patrick Levallois, Suzanne Gingras, Sylvie Marcoux, Christelle Legay, Cyril Catto, Manuel Rodriguez, Robert Tardif
ABSTRACT
There is concern about possible effects of disinfection by-products on reproductive outcomes. The purpose of this study was to evaluate the association between maternal exposure to chlorination by-products and the risk of delivering a small for-gestational-age (SGA) neonate. We conducted a population-based case-control study in the Québec City (Canada) area. Term newborn cases with birth weights <10th percentile (n = 571) were compared with 1925 term controls with birth weights ≥10th percentile. Concentrations of trihalomethanes and haloacetic acids in the water-distribution systems of participants were monitored during the study period, and a phone interview on maternal habits was completed within 3 months after childbirth. We estimated chlorination by-products ingestion during the last trimester of pregnancy and trihalomethanes doses resulting from inhalation and dermal exposure. We evaluated associations between chlorination by-products in utero exposure and SGA by means of unconditional logistic regression with control of potential confounders. When total trihalomethanes and the 5 regulated haloacetic acids concentrations were divided into quartiles, no clear dose-response relationship was found with SGA. However, increased risk was observed when haloacetic concentrations were above the fourth quartile and when either trihalomethanes or haloacetic acids concentrations were above current water standards (adjusted OR= 1.5 [95% confidence interval = 1.1-1.9] and 1.4 [1.1-1.9], respectively). Inhalation and dermal absorption of trihalomethanes did not contribute to this risk, but a monotonic dose-response was found with haloacetic acids ingestion. Oral exposure to high levels of chlorination by-products in drinking water could be a risk factor for term SGA.
Product Description
Supelco
Chloroacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Chloroacetic acid, 99%
Sigma-Aldrich
Chloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloroacetate, 98%