MilliporeSigma
  • Dual-functional gadolinium-based copper(II) probe for selective magnetic resonance imaging and fluorescence sensing.
Dual-functional gadolinium-based copper(II) probe for selective magnetic resonance imaging and fluorescence sensing.
Inorganic chemistry (2012-02-10)
Xiaolin Zhang, Xu Jing, Tao Liu, Gang Han, Huaqiang Li, Chunying Duan
ABSTRACT
A unique gadolinium complex, Nap-DO3A-Gd, comprising a naphthylamine luminescent moiety, a di-2-picolylamine (DPA) binding chelator, and a 1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid (DO3A) moiety has been designed and synthesized as a dual-functional probe for selective magnetic resonance imaging and fluorescent sensing of copper(II) in living cells. Nap-DO3A-Gd exhibited a turn-on manner of relaxivity changes and a fluorescent quenching toward Cu(2+). Through the introduction of naphthalamide into the Gd(3+) contrast agent platform to restrict the coordination ability of the DPA chelator and with Gd(3+) coordinating to the DPA moiety to turn away the interferences of other metal cations from Cu(2+) detection, the probe featured selective relaxivity changes toward Cu(2+) over other metal ions and brought unique Cu(2+)-specific luminescent responses. The probe was water-soluble with the luminescent detection limit established at 6 ppb and was successfully used for luminescence imaging detection of copper(II) in living cells. The results demonstrated the efficiency and advantage of our approach in the development of a dual-modality image.
Product Description
Millipore
Nitrate Reagent A, suitable for microbiology
Supelco
1-Naphthylamine, analytical standard
Sigma-Aldrich
1-Naphthylamine, 97%