MilliporeSigma
2-Aminopyrimidine as a novel scaffold for biofilm modulation.
Organic & biomolecular chemistry (2012-02-04)
Erick A Lindsey, Roberta J Worthington, Cristina Alcaraz, Christian Melander
ABSTRACT
An efficient synthetic route to a series of substituted 2-aminopyrimidine (2-AP) derivatives has been developed. Subsequent biofilm screening has allowed comparison between the biological activity of these new derivatives and that of the 2-aminoimidazole class of anti-biofilm compounds. Several derivatives displayed the ability to modulate bacterial biofilm formation, exhibiting greater activity against Gram-positive strains than Gram-negative strains. Additionally some 2-aminopyrmidines were able to suppress MRSA resistance to conventional antibiotics.
Sigma-Aldrich
2-Aminopyrimidine, 97%