MilliporeSigma
  • Convergent integration of two self-labor domino sequences: a novel method for the synthesis of oxazole derivatives from methyl ketones and benzoins.
Convergent integration of two self-labor domino sequences: a novel method for the synthesis of oxazole derivatives from methyl ketones and benzoins.
Chemical communications (Cambridge, England) (2012-01-31)
Wei-Jian Xue, Qi Li, Yan-Ping Zhu, Jun-Gang Wang, An-Xin Wu
ABSTRACT
A highly efficient method for the synthesis of oxazole derivatives from methyl ketones, benzoins and ammonium acetate has been established via a novel strategy-convergent integration of two self-labor domino sequences. Owing to the simple and readily available starting materials, mild reaction conditions, facile operation, and the high bioactivity of oxazole derivatives, this reaction promises diverse applications in medical chemistry. Additionally, this reaction could provide an efficient example for self-labor synthesis strategy of organic compounds.
Sigma-Aldrich
Benzoin, 98%
Sigma-Aldrich
Benzoin, purified by sublimation, ≥99.5%