MilliporeSigma
Amplified microRNA detection by templated chemistry.
Nucleic acids research (2012-01-27)
Emily M Harcourt, Eric T Kool
ABSTRACT
MicroRNAs (miRNAs) are a class of RNAs that play important regulatory roles in the cell. The detection of microRNA has attracted significant interest recently, as abnormal miRNA expression has been linked to cancer and other diseases. Here, we present a straightforward method for isothermal amplified detection of miRNA that involves two separate nucleic acid-templated chemistry steps. The miRNA first templates the cyclization of an oligodeoxynucleotide from a linear precursor containing a 5'-iodide and a 3'-phosphorothioate. The sequence is amplified through rolling circle amplification with 29 DNA polymerase and then detected via a second amplification using fluorogenic templated probes. Tests showed that the cyclization proceeds in ∼50% yield over 24 h and is compatible with the conditions required for rolling circle polymerization, unlike enzymatic ligations which required non-compatible buffer conditions. The polymerization yielded 188-fold amplification, and separate experiments showed ∼15-fold signal amplification from the templated fluorogenic probes. When all components are combined, results show miRNA detection down to 200 pM in solution, and correlation of the detected signal with the initial concentration of miRNA. The doubly templated double-amplification method demonstrates a new approach to detection of rolling circle products and significant advantages in ease of operation for miRNA detection.
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)