MilliporeSigma
  • Efficient siRNA delivery using a polyamidoamine dendrimer with a modified pentaerythritol core.
Efficient siRNA delivery using a polyamidoamine dendrimer with a modified pentaerythritol core.
Pharmaceutical research (2012-01-26)
Yue Zhang, Chenguang Zhou, Kwang Joo Kwak, Xinmei Wang, Bryant Yung, L James Lee, Yanming Wang, Peng George Wang, Robert J Lee
ABSTRACT
Delivery of siRNA into cells remains a critical challenge. Our lab has shown a novel polyamidoamine (PAMAM) dendrimer with modified pentaerythritol derivative core (PD dendrimer) to exhibit high plasmid DNA transfection efficiency and low cytotoxicity. Here, we evaluate PD dendrimer as a siRNA carrier. Agarose gel electrophoresis and AFM were used to confirm formation of generation 5 (G5)-PD dendrimer/siRNA nanoparticles (NPs). G5 PD dendrimer/anti-luciferase siRNA NPs were used to transfect SK Hep-1 cells with stable luciferase expression. Effects of various endocytic pathway inhibitors on uptake of G5 PD dendrimer/siRNA NPs in SK Hep-1 cells were also investigated. Agarose gel electrophoresis indicated that G5 PD dendrimer and siRNA formed NPs at weight ratios >0.5:1. G5 PD dendrimer showed effective luciferase gene silencing when weight ratio was 3.0:1 and above. Treatment with endocytosis inhibitors showed that clathrin-mediated endocytosis was the main endocytic pathway by which G5-PD dendrimer/siRNA NPs enter the cell. These results show that the novel G5 PD dendrimer has high siRNA delivery activity and is promising as a delivery agent for its therapeutic application.
Sigma-Aldrich
Pentaerythritol, 99%
Sigma-Aldrich
Pentaerythritol, 98%