MilliporeSigma
  • A novel sensitive DNA-biosensor for detection of a carcinogen, Sudan II, using electrochemically treated pencil graphite electrode by voltammetric methods.
A novel sensitive DNA-biosensor for detection of a carcinogen, Sudan II, using electrochemically treated pencil graphite electrode by voltammetric methods.
Talanta (2012-01-24)
Ali A Ensafi, Behzad Rezaei, Maryam Amini, Esmaeil Heydari-Bafrooei
ABSTRACT
A simple and inexpensive methodology was used to develop a novel electrochemical sensor for the determination of Sudan II. The interaction of Sudan II with salmon sperm ds-DNA on the surface of salmon sperm ds-DNA-modified pencil graphite electrode (PGE) and in solution phase was studied, using differential pulse voltammetry. The difference between adenine and guanine signals of the ds-DNA after and before interaction with Sudan II was directly proportional to Sudan II concentration, which used for quantitative inspections. Using PGE, a linear calibration curve (R(2)=0.9958) was observed with 0.5-6.0 μg mL(-1) Sudan II. Furthermore, the LOD of 0.4 μg mL(-1) and linear range between 0.5 and 4.0 μg mL(-1) were achieved in solution phase. In the second part, Sudan II was determined on a pretreated pencil graphite electrode by means of adsorptive stripping differential pulse voltammetry. The peak current was linearly dependent on Sudan II concentration over the range of 0.0015-0.30 μg mL(-1), with a detection limit of 0.00007 μg mL(-1) Sudan II. Both ds-DNA-modified PGE and PPGE were applied to analyze Sudan II in real samples.
Product Description
Supelco
Sudan II, analytical standard
Sigma-Aldrich
Sudan II, Dye content 90 %