MilliporeSigma
  • Release of flavonoids from lupin globulin proteins during digestion in a model system.
Release of flavonoids from lupin globulin proteins during digestion in a model system.
Journal of agricultural and food chemistry (2012-01-24)
Jaroslaw Czubinski, Krzysztof Dwiecki, Aleksander Siger, Piotr Kachlicki, Grazyna Neunert, Eleonora Lampart-Szczapa, Malgorzata Nogala-Kalucka
ABSTRACT
Lupin seed globulin proteins form complexes with flavonoids, predominantly apigenin C-glycosides. Enzymes typical for the gastrointestinal tract were used to hydrolyze lupin seed globulins. Release of native flavonoids as a result of the proteolysis reaction was observed. Different analytical methods such as size exclusion chromatography, HPLC-MS, and fluorescence spectroscopy (steady-state fluorescence, fluorescence anisotropy, fluorescence lifetimes) were used for a detailed characterization of this phenomenon. Flavonoids liberated from lupin globulin proteins as a result of pancreatin-catalyzed digestion were bound by γ-conglutin resistant to this enzyme. Two possible mechanisms of this interaction may be suggested: hydrogen bonding between oligosaccharide chains of glycoproteins and the sugar moieties of the flavonoid glycosides or electrostatic attraction between positively charged γ-conglutin and flavonoids partially ionized at pH 7.5.
Sigma-Aldrich
Pancreatin from porcine pancreas, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Sigma-Aldrich
Pancreatin from porcine pancreas, ≥3 × USP specifications
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications