MilliporeSigma
  • [Effects of seed soaking with humic acid on wheat seedlings antioxidant system under salt-alkali stress].
[Effects of seed soaking with humic acid on wheat seedlings antioxidant system under salt-alkali stress].
Ying yong sheng tai xue bao = The journal of applied ecology (2012-01-24)
Wei Guo, Qing-xiang Wang
ABSTRACT
The seeds of two spring wheat genotypes Longmai 26 and Kehan 16 were soaked with humic acid to study the effects of seed soaking with humic acid on the seedlings antioxidant system of the two genotypes under NaCl and Na2CO3 stresses. Under the salt and alkali stresses, the seedlings leaf proline (Pro) content increased, membrane permeability enhanced, and aboveground fresh mass decreased. Under NaCl stress, the leaf catalase (CAT) activity increased, peroxidase (POD) activity decreased, and superoxide dismutase (SOD) decreased after an initial increase; under Na2CO3 stress, the CAT and POD activities increased while the SOD activity was in adverse. The activities of the test antioxidant enzymes were higher under Na2CO3 than under NaCl stress. When the seeds were soaked with humic acid, the seedlings leaf glutathione (GSH) content increased, and the SOD and CAT activities improved, which effectively relieved the damage of salt-alkali stress on the seedlings growth. Some differences were observed in the resistance level and related mechanisms against the salt-alkali stress between the two wheat genotypes, possibly due to their different interactions with the salt ion components and concentrations and the ways of seed soaking.
Product Description
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate decahydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate monohydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate monohydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate monohydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7