MilliporeSigma
  • Excited state electric dipole moment of nile blue and brilliant cresyl blue: a comparative study.
Excited state electric dipole moment of nile blue and brilliant cresyl blue: a comparative study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-01-21)
A Ghanadzadeh Gilani, S E Hosseini, M Moghadam, E Alizadeh
ABSTRACT
A solvatochromic study on the photophysical properties of two cationic oxazine dyes (brilliant cresyl blue and nile blue) was carried out. The electronic absorption and emission spectra of the dyes were recorded in various organic solvents with different polarity. The ground and the excited state dipole moments of the dyes were estimated from solvatochromic shift method. The solvent dependent spectral shifts in absorption and fluorescence spectra were analyzed by the Katritzky and Kamlet-Taft multi-parameter scales. This work is characterized by detailed quantitative studies on the nature and extent of solvent-solute interactions.
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %
Pricing and availability is not currently available.
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %
Pricing and availability is not currently available.
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Pricing and availability is not currently available.
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %
Pricing and availability is not currently available.