MilliporeSigma
  • A comprehensive review of intake estimates of di-isononyl phthalate (DINP) based on indirect exposure models and urinary biomonitoring data.
A comprehensive review of intake estimates of di-isononyl phthalate (DINP) based on indirect exposure models and urinary biomonitoring data.
Regulatory toxicology and pharmacology : RTP (2012-01-19)
Kevin M Kransler, Ammie N Bachman, Richard H McKee
ABSTRACT
Di-isononyl phthalate (DINP) is a high molecular weight general purpose plasticizer used principally in the manufacture of flexible polyvinyl chloride (PVC) articles. DINP metabolites can be measured in biological media such as blood and urine. However, measurement of a substance in the blood or urine does not by itself mean that the chemical causes or is associated with adverse health outcomes. This is particularly pertinent given the advances in modern analytical techniques whereby ever diminishing trace amounts of substances can be detected. Therefore, it is a scientific necessity that risk assessors understand the relationship of biomonitoring data to estimation of exposure so that appropriate comparisons can be made to the no observed adverse effects levels (NOAELs) or other points of departure from toxicological studies in animals. In this paper, estimates of daily DINP intake are calculated for various population segments based on urinary biomonitoring data and are compared to estimates of exposure based on indirect methods and to health-based exposure guidance values. In general, intake estimates converge on a mean of 1-2μg/kg/day regardless of source of exposure or population cluster; a value 2-orders of magnitude lower than health-based exposure guidance values, ranging from 120 to 290μg/kg/day, which have been established by regulatory authorities and other authoritative bodies as representing acceptable levels.
Sigma-Aldrich
Diisononyl phthalate, ester content ≥99 % (mixture of C9 isomers), technical grade
Pricing and availability is not currently available.
Supelco
Bis(7-methyloctyl) phthalate, analytical standard
Pricing and availability is not currently available.