MilliporeSigma
  • Erasure of a spinal memory trace of pain by a brief, high-dose opioid administration.
Erasure of a spinal memory trace of pain by a brief, high-dose opioid administration.
Science (New York, N.Y.) (2012-01-17)
Ruth Drdla-Schutting, Justus Benrath, Gabriele Wunderbaldinger, Jürgen Sandkühler
ABSTRACT
Painful stimuli activate nociceptive C fibers and induce synaptic long-term potentiation (LTP) at their spinal terminals. LTP at C-fiber synapses represents a cellular model for pain amplification (hyperalgesia) and for a memory trace of pain. μ-Opioid receptor agonists exert a powerful but reversible depression at C-fiber synapses that renders the continuous application of low opioid doses the gold standard in pain therapy. We discovered that brief application of a high opioid dose reversed various forms of activity-dependent LTP at C-fiber synapses. Depotentiation involved Ca(2+)-dependent signaling and normalization of the phosphorylation state of α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptors. This also reversed hyperalgesia in behaving animals. Opioids thus not only temporarily dampen pain but may also erase a spinal memory trace of pain.
Sigma-Aldrich
CTOP, ≥97% (HPLC)