MilliporeSigma
  • Bioassay-guided isolation of an alkaloid with antiangiogenic and antitumor activities from the extract of Fissistigma cavaleriei root.
Bioassay-guided isolation of an alkaloid with antiangiogenic and antitumor activities from the extract of Fissistigma cavaleriei root.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2012-01-14)
Zaichang Yang, Weilun Lu, Xiaoyan Ma, Dandan Song
ABSTRACT
Fissistigma cavaleriei (Levl) Rehd (Annonaceae) is used as a folklore medicine for treatment of inflammation, arthritis, and tuberculosis by Miao people in China. In the present study, the antiangiogenic activity of F. cavaleriei was investigated. The chorioallantoic membrane of the fertilized hen's egg (CAM assay) was used to determine antiangiogenic activity of the plant extract. Compound (1), a compound with antiangiogenic activity, was isolated by bioassay-guided fractionation from F. cavaleriei for the first time. The structure of compound (1) was elucidated on the basis of spectroscopic methods. Colorimetric COX (ovine) inhibitor screening assay was used to determine its inhibitory effect on COX-1 and COX-2. MTT and Sulforhodamine B assays were used to investigate its cytotoxic effects on tumor cell lines. As a result, compound (1) showed a selectively inhibiting effect on COX-2 and could inhibit the growth of tumor cells in vitro. The antitumor activity of compound (1) was further confirmed by the observation that compound (1) administration significantly inhibited the growth of S-180 cells in mice. Moreover, compound (1) was able to enhance the antitumor activity of doxorubicin in the mice bearing with S-180 cells while combined with doxorubicin. In conclusion, compound (1) is a multi-target molecule and further experimental investigations are needed to determine whether it can be used as a lead molecule for tumor treatment.
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sulforhodamine B, acid form, Dye content 95 %
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture