MilliporeSigma
  • Peptide-mediated nanoengineering of inorganic particle surfaces: a general route toward surface functionalization via peptide adhesion domains.
Peptide-mediated nanoengineering of inorganic particle surfaces: a general route toward surface functionalization via peptide adhesion domains.
Journal of the American Chemical Society (2012-01-14)
Thorsten Schwemmer, Jens Baumgartner, Damien Faivre, Hans G Börner
ABSTRACT
The peptide-mediated functionalization of inorganic particle surfaces is demonstrated on gadolinium oxide (GdO) particles, revealing specific means to functionalize nano- or microparticles. Phage display screening is exploited to select 12mer peptides, which exhibit sequence-specific adhesion onto surfaces of GdO particles. These peptide adhesion domains are exploited to effectively decorate GdO particles with fluorescently labeled poly(ethylene oxide) (PEO), proving to result in a stable surface modification as shown by significant reduction of protein adsorption by 80%, compared to nonfunctionalized particles. Peptide adhesion and stability of the noncovalent coating are investigated by adsorption/elution experiments and Langmuir isotherms. Fluorescence microscopy, contact angle, and energy dispersive X-ray (EDX) measurements confirmed the sequence specificity of the interactions by comparing adhesion sequences with scrambled peptide sequences. Noncovalent, but specific modification of inorganic particle surfaces represents a generic strategy to modulate functionality and function of nano- or microparticle surfaces.
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis