MilliporeSigma
  • Possibilities of regulating disorders in functional activity of the retina with antioxidants under conditions of high-intensity light exposure.
Possibilities of regulating disorders in functional activity of the retina with antioxidants under conditions of high-intensity light exposure.
Bulletin of experimental biology and medicine (2012-01-12)
Sh Y Mamedov, E M Kasimov, A I Djafarov
ABSTRACT
We studied the effects of natural (α-tocopherol and SOD) and synthetic (potassium phenosane and hydroxypyridine) antioxidants on the recovery of disordered parameters of the electroretinogram under conditions of high-intensity light exposure. Potassium phenosane and tocopherol acetate more effectively normalized parameters of electroretinogram in monotherapy. The use of a complex of antioxidants completely prevented the disorders in the electroretinogram parameters in retinopathy after exposure to intense light.
Sigma-Aldrich
Phenothiazine, purum, ≥98.0% (GC)
Sigma-Aldrich
Phenothiazine, ≥98%