MilliporeSigma
  • Assessment of enveloping distribution sampling to calculate relative free enthalpies of binding for eight netropsin-DNA duplex complexes in aqueous solution.
Assessment of enveloping distribution sampling to calculate relative free enthalpies of binding for eight netropsin-DNA duplex complexes in aqueous solution.
Journal of computational chemistry (2012-01-10)
Niels Hansen, Jožica Dolenc, Matthias Knecht, Sereina Riniker, Wilfred F van Gunsteren
ABSTRACT
The performance of enveloping distribution sampling (EDS) simulations to estimate free enthalpy differences associated with seven alchemical transformations of A-T into G-C base pairs at the netropsin binding site in the minor groove of a 13-base pair DNA duplex in aqueous solution is evaluated. It is demonstrated that sufficient sampling can be achieved with a two-state EDS Hamiltonian even for large perturbations such as the simultaneous transformation of up to three A-T into three G-C base pairs. The two parameters required to define the EDS reference state Hamiltonian are obtained automatically using a modified version of a scheme presented in earlier work. The sensitivity of the configurational sampling to a variation of these parameters is investigated in detail. Although for relatively small perturbations, that is, one base pair, the free enthalpy estimate depends only weakly on the EDS parameters, the sensitivity is stronger for the largest perturbation. Yet, EDS offers various convenient measures to evaluate the degree of sampling and thus the reliability of the free enthalpy estimate and appears to be an efficient alternative to the conventional thermodynamic integration methodology to obtain free energy differences for molecular systems.
Sigma-Aldrich
Netropsin dihydrochloride, from Streptomyces netropsis, ≥98% (HPLC), powder