MilliporeSigma
  • Metabolomic profiling of the flower bud and rachis of Tussilago farfara with antitussive and expectorant effects on mice.
Metabolomic profiling of the flower bud and rachis of Tussilago farfara with antitussive and expectorant effects on mice.
Journal of ethnopharmacology (2012-01-03)
Zhen-Yu Li, Hai-Juan Zhi, Shui-Yu Xue, Hai-Feng Sun, Fu-Sheng Zhang, Jin-Ping Jia, Jie Xing, Li-Zeng Zhang, Xue-Mei Qin
ABSTRACT
Flower bud of Tussilago farfara L. is widely used for the treatment of cough, bronchitis and asthmatic disorders in the Traditional Chinese Medicine. However, due to the increasing demands, adulteration with rachis is frequently encountered in the marketplace. No report demonstrated the chemical and pharmacological differences between flower bud and rachis before. The water extracts were orally administrated to mice. Ammonia induced mice coughing model was used to evaluate the antitussive activity. The expectorant activity was evaluated by volume of phenol red in mice's tracheas. Metabolites were identified directly from the crude extracts through 1D- and 2D-NMR spectra. A metabolic profiling carried out by (1)H NMR spectroscopy and multivariate data analysis was applied to crude extracts from flower bud and rachis. Flower bud significantly lengthened the latent period of cough, decreased cough frequency caused by ammonia and enhanced tracheal phenol red output in expectorant evaluation. Principal component analysis (PCA) yielded good separation between flower bud and rachis, and corresponding loading plot showed that the phenolic compounds, organic acid, sugar, amino acid, terpene and sterol contributed to the discrimination. These findings provide pharmacological and chemical evidence that only flower bud can be used as the antitussive and expectorant herbal drug. The high concentration of chlorogenic acid, 3,5-dicaffeoylquinic acid, rutin in flower buds may be related with the antitussive and expectorant effects of Flos Farfara. To guarantee the clinical effect, rachis should be picked out before use.
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture