MilliporeSigma
  • Verbascoside is not genotoxic in the ST and HB crosses of the Drosophila wing spot test, and its constituent, caffeic acid, decreases the spontaneous mutation rate in the ST cross.
Verbascoside is not genotoxic in the ST and HB crosses of the Drosophila wing spot test, and its constituent, caffeic acid, decreases the spontaneous mutation rate in the ST cross.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-12-27)
Luis Felipe Santos-Cruz, José Guillermo Ávila-Acevedo, Diego Ortega-Capitaine, Jesús Clemente Ojeda-Duplancher, Juana Laura Perdigón-Moya, Luis Barbo Hernández-Portilla, Héctor López-Dionicio, Angel Durán-Díaz, Irma Elena Dueñas-García, Laura Castañeda-Partida, Ana María García-Bores, María Eugenia Heres-Pulido
ABSTRACT
Verbascoside (VB) is a phenylpropanoid isolated from Buddleja species, some of which originate in Mexico, and was first described in the sixteenth century in the codices of Mexican traditional medicine. VB is present in alcohol extracts and is widely used in the north of Mexico as a sunscreen. VB absorbs UV-A and UV-B radiation and has high antioxidant and anti-inflammatory capacities. VB and its constituent caffeic acid (CA) were screened to determine their genotoxic activity using the Drosophila wing spot test. Third instar larvae (72±4 h) of the standard (ST) and high bioactivation (HB) crosses, with regulated and high levels of cytochrome P450s (Cyp450s), respectively, were exposed to VB or CA (0, 27, 57, 81, 135, and 173 mM). VB was not genotoxic at any of the concentrations tested in both crosses. The amount of VB residue as determined by HPLC in the adult flies that were fed with VB indicated a low metabolism of this compound, which explains the absence of genotoxicity. CA decreased the spontaneous frequencies of small and total spots and showed putative toxicity in the ST cross.
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)
Verbascoside, primary reference standard