MilliporeSigma
  • Efficacy and safety of doxepin 6 mg in a four-week outpatient trial of elderly adults with chronic primary insomnia.
Efficacy and safety of doxepin 6 mg in a four-week outpatient trial of elderly adults with chronic primary insomnia.
Sleep medicine (2011-12-27)
Alan Lankford, Roberta Rogowski, Beal Essink, Elizabeth Ludington, H Heith Durrence, Thomas Roth
ABSTRACT
The efficacy and safety of doxepin (DXP), a histamine H(1) receptor antagonist, was evaluated in elderly adults with sleep maintenance insomnia. This was a randomized, double-blind, placebo-controlled outpatient trial. Elderly adults meeting DSM-IV-TR criteria for primary insomnia were randomized to four weeks of nightly treatment with either DXP 6 mg (N=130) or placebo (PBO; N=124). Efficacy was assessed using patient self-report instruments and clinician ratings. Patient-reported endpoints included subjective total sleep time (sTST), subjective wake after sleep onset (sWASO), latency to sleep onset (LSO), sleep quality, and a Patient Global Impression scale (PGI). The primary endpoint was sTST at week 1. DXP 6 mg produced significantly more sTST and less sWASO at week 1 (both p-values <0.0001) than PBO. These significant improvements versus placebo were maintained at weeks 2-4 (all p-values <0.05). There were no significant differences in LSO for DXP 6 mg versus PBO. DXP 6 mg significantly improved sleep quality (weeks 1, 3, and 4, p<0.05) and several outcome-related parameters, including several items on the PGI, the severity and improvement items of the Clinician Global Impression scale (CGI; weeks 1 and 2) and the Insomnia Severity Index (ISI; weeks 1-4), all versus PBO. There were no reports of anticholinergic effects (e.g., dry mouth) or memory impairment. The safety profile of DXP 6 mg was comparable to that of PBO. In elderly adults with insomnia, DXP 6 mg produced significant improvements in sleep maintenance, sleep duration, and sleep quality endpoints that were sustained throughout the trial. These data suggest that DXP 6 mg is effective for treating sleep maintenance insomnia and is well-tolerated in elderly adults with chronic primary insomnia.
Sigma-Aldrich
Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder
Doxepin for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Doxepin hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®