MilliporeSigma
  • An inorganic-organic hybrid optical sensor for heavy metal ion detection based on immobilizing 4-(2-pyridylazo)-resorcinol on functionalized HMS.
An inorganic-organic hybrid optical sensor for heavy metal ion detection based on immobilizing 4-(2-pyridylazo)-resorcinol on functionalized HMS.
Journal of hazardous materials (2011-12-20)
Tongge Liu, Gang Li, Na Zhang, Yongying Chen
ABSTRACT
A novel and low-cost optical sensor for heavy metal ion detection has been prepared by immobilizing 4-(2-pyridylazo)-resorcinol (PAR) on the functionalized hexagonal mesoporous silica (HMS) via N-trimethoxysilylpropyl-N,N,N-trimethylammonium chloride (TMAC). The successful fabrication of this optical sensor is confirmed by extensive characterizations using FT-IR, low angle X-ray diffraction (XRD), UV-vis spectroscopy and N(2) sorption, meanwhile its colorimetric properties, selectivity, sensitivity and reversibility are also investigated. The optical sensor responds selectively to heavy metal ions, such as Fe(3+), Cd(2+), Ni(2+), Zn(2+), Pb(2+), Co(2+), Hg(2+) and Cu(2+) with a color change from yellow to orange or purple in alkaline solutions, while it shows a color change only for Cu(2+) under strongly acidic conditions. At pH 12.0, this optical sensor has a high sensitivity that makes it possible to detect Cu(2+) in aqueous solution with the detection limit as low as 40ppb by naked-eye. This optical sensor also shows excellent reversibility and regeneration by treatment with a solution of EDTA.
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
4-(2-Pyridylazo)resorcinol, 96%