MilliporeSigma
Comparison of different treatment modalities for oral halitosis.
Acta odontologica Scandinavica (2011-12-14)
Seida Erovic Ademovski, Peter Lingström, Edwin Winkel, Albert Tangerman, G Rutger Persson, Stefan Renvert
ABSTRACT
To assess the effects on intra-oral halitosis by a mouth rinse containing zinc acetate (0.3%) and chlorhexidine diacetate (0.025%) with and without adjunct tongue scraping. Twenty-one subjects without a diagnosis of periodontitis were randomized in a cross-over clinical trial. Organoleptic scores (OLS) were assessed to define intra-oral halitosis by total volatile sulfur compound (T-VSC) measurements and by gas chromatography. Twenty-one subjects with a mean age of 45.7 years (SD: ±13.3, range: 21-66). The OLS were significantly lower following active rinse combined with tongue scraping (p < 0.001) at all time points. Immediately after, at 30 min, and at day 14, the T-VSC values were lower in the active rinse sequence than in the negative rinse sequence (p < 0.001, p < 0.001 and p < 0.05, respectively). At 30 min and at day 14, the hydrogen sulfide (H(2)S) and methyl mercaptan (MM) values were lower in the active rinse sequence compared to the inactive rinse sequence (p < 0.001). The inactive rinse sequence with tongue scraping reduced T-VSC at 30 min (p < 0.001) but not at 14 days. Similar reductions in T-VSC, H(2)S and MM were found in the active rinse sequence with or without tongue scraping. The use of a tongue scraper did not provide additional benefits to the active mouth rinse, but reduced OLS and tongue coating index.
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%