Merck
  • Covalent immobilization of catalase onto spacer-arm attached modified florisil: characterization and application to batch and plug-flow type reactor systems.
Covalent immobilization of catalase onto spacer-arm attached modified florisil: characterization and application to batch and plug-flow type reactor systems.
Enzyme and microbial technology (2011-12-07)
Ozlem Alptekin, S Seyhan Tükel, Deniz Yildirim, Dilek Alagöz
ABSTRACT
Catalase was covalently immobilized onto florisil via glutaraldehyde (GA) and glutaraldehyde+6-amino hexanoic acid (6-AHA) (as a spacer arm). Immobilizations of catalase onto modified supports were optimized to improve the efficiency of the overall immobilization procedures. The V(max) values of catalase immobilized via glutaraldehyde (CIG) and catalase immobilized via glutaraldehyde+6-amino hexanoic acid (CIG-6-AHA) were about 0.6 and 3.4% of free catalase, respectively. The usage of 6-AHA as a spacer arm caused about 40 folds increase in catalytic efficiency of CIG-6-AHA (8.3 × 10⁵ M⁻¹ s⁻¹) as compared to that of CIG (2.1 × 10⁴ M⁻¹ s⁻¹). CIG and CIG-6-AHA retained 67 and 35% of their initial activities at 5 °C and 71 and 18% of their initial activities, respectively at room temperature at the end of 6 days. Operational stabilities of CIG and CIG-6-AHA were investigated in batch and plug-flow type reactors. The highest total amount of decomposed hydrogen peroxide (TAD-H₂O₂) was determined as 219.5 μmol for CIG-6-AHA in plug-flow type reactor.
Product Description
Supelco
Florisil® Adsorbent for Chromatography, 60-100 mesh
Supelco
Florisil® Adsorbent for Chromatography, 30-60 mesh
Supelco
Florisil®, 100-200 mesh, fine powder
Supelco
Florisil®, PR grade, 60-100 mesh, coarse powder
Supelco
Florisil®, for the determination of hydrocarbon acc. to ISO 9377-2
Supelco
Florisil®, <200 mesh, fine powder
Supelco
Florisil® PR, PR grade (analysis acc. to FDA)
Sigma-Aldrich
Florisil®, 100-200 mesh