MilliporeSigma
  • Classification of smoke tainted wines using mid-infrared spectroscopy and chemometrics.
Classification of smoke tainted wines using mid-infrared spectroscopy and chemometrics.
Journal of agricultural and food chemistry (2011-12-02)
Anthea L Fudge, Kerry L Wilkinson, Renata Ristic, Daniel Cozzolino
ABSTRACT
In this study, the suitability of mid-infrared (MIR) spectroscopy, combined with principal component analysis (PCA) and linear discriminant analysis (LDA), was evaluated as a rapid analytical technique to identify smoke tainted wines. Control (i.e., unsmoked) and smoke-affected wines (260 in total) from experimental and commercial sources were analyzed by MIR spectroscopy and chemometrics. The concentrations of guaiacol and 4-methylguaiacol were also determined using gas chromatography-mass spectrometry (GC-MS), as markers of smoke taint. LDA models correctly classified 61% of control wines and 70% of smoke-affected wines. Classification rates were found to be influenced by the extent of smoke taint (based on GC-MS and informal sensory assessment), as well as qualitative differences in wine composition due to grape variety and oak maturation. Overall, the potential application of MIR spectroscopy combined with chemometrics as a rapid analytical technique for screening smoke-affected wines was demonstrated.
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%, FG
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%
Sigma-Aldrich
2-Methoxy-4-methylphenol, natural, 97%, FG