MilliporeSigma
  • Triterpene functional genomics in licorice for identification of CYP72A154 involved in the biosynthesis of glycyrrhizin.
Triterpene functional genomics in licorice for identification of CYP72A154 involved in the biosynthesis of glycyrrhizin.
The Plant cell (2011-12-01)
Hikaru Seki, Satoru Sawai, Kiyoshi Ohyama, Masaharu Mizutani, Toshiyuki Ohnishi, Hiroshi Sudo, Ery Odette Fukushima, Tomoyoshi Akashi, Toshio Aoki, Kazuki Saito, Toshiya Muranaka
ABSTRACT
Glycyrrhizin, a triterpenoid saponin derived from the underground parts of Glycyrrhiza plants (licorice), has several pharmacological activities and is also used worldwide as a natural sweetener. The biosynthesis of glycyrrhizin involves the initial cyclization of 2,3-oxidosqualene to the triterpene skeleton β-amyrin, followed by a series of oxidative reactions at positions C-11 and C-30, and glycosyl transfers to the C-3 hydroxyl group. We previously reported the identification of a cytochrome P450 monooxygenase (P450) gene encoding β-amyrin 11-oxidase (CYP88D6) as the initial P450 gene in glycyrrhizin biosynthesis. In this study, a second relevant P450 (CYP72A154) was identified and shown to be responsible for C-30 oxidation in the glycyrrhizin pathway. CYP72A154 expressed in an engineered yeast strain that endogenously produces 11-oxo-β-amyrin (a possible biosynthetic intermediate between β-amyrin and glycyrrhizin) catalyzed three sequential oxidation steps at C-30 of 11-oxo-β-amyrin supplied in situ to produce glycyrrhetinic acid, a glycyrrhizin aglycone. Furthermore, CYP72A63 of Medicago truncatula, which has high sequence similarity to CYP72A154, was able to catalyze C-30 oxidation of β-amyrin. These results reveal a function of CYP72A subfamily proteins as triterpene-oxidizing enzymes and provide a genetic tool for engineering the production of glycyrrhizin.
Product Description
Supelco
β-Amyrin, analytical standard
Supelco
α-Amyrin, analytical standard