MilliporeSigma
  • Effect of dietary vanadium on the ileac T cells and contents of cytokines in broilers.
Effect of dietary vanadium on the ileac T cells and contents of cytokines in broilers.
Biological trace element research (2011-11-26)
Kangping Wang, Hengmin Cui, Yuanxin Deng, Xi Peng, Jing Fang, Zhicai Zuo, Wei Cui
ABSTRACT
The purpose of this 42-day study was to examine the effect of dietary vanadium on the ileac T cells and contents of cytokines including interleukin-2 (IL-2), interleukin-6 (IL-6), and interferon-gamma (IFN-γ) in broilers by flow cytometry and enzyme-linked immunosorbent assay. A total of 420 one-day-old avian broilers were divided into six groups (seven replicates in each group and ten broilers in each replicate) and fed on control diet or the same diet supplemented with 5, 15, 30, 45, and 60 mg/kg vanadium in the form of ammonium metavanadate. The results showed that the percentages of CD3(+), CD3(+)CD4(+), and CD3(+)CD8(+) T cells in both ileac lamina propria lymphocytes (LPLs) and intraepithelial lymphocytes (IELs) were significantly lower (P < 0.05 or P < 0.01) in the 45- and 60-mg/kg groups than in the control group from 14 to 42 days of age. The CD4(+)/CD8(+) ratio was increased in ileac LPLs in the 60-mg/kg group at 28 days of age, and in ileac IELs in the 60-mg/kg group at 28 days of age and in the 45-mg/kg group at 42 days of age. Meanwhile, the ileac IL-2, IL-6 contents were decreased (P < 0.05 or P < 0.01) in the 60-mg/kg group from 14 to 42 days of age and in the 45-mg/kg group from 28 to 42 days of age in comparison with those of the control group. It was concluded that dietary vanadium in excess of 30 mg/kg reduced the ileac T cell population and percentages of T cell subsets, and IL-2, IL-6, and IFN-γ contents, implying that the immune function of local intestinal mucosa in broilers could be affected by the dietary vanadium.
Sigma-Aldrich
Ammonium metavanadate, 99.95% trace metals basis
Sigma-Aldrich
Ammonium metavanadate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Ammonium metavanadate, 99%
Sigma-Aldrich
Ammonium metavanadate, ACS reagent, ≥99.0%