MilliporeSigma
  • Lipase-catalyzed kinetic resolution of aryltrimethylsilyl chiral alcohols.
Lipase-catalyzed kinetic resolution of aryltrimethylsilyl chiral alcohols.
Molecules (Basel, Switzerland) (2011-11-25)
Dayvson J Palmeira, Juliana C Abreu, Leandro H Andrade
ABSTRACT
Lipase-catalyzed kinetic resolution of aryltrimethylsilyl chiral alcohols through a transesterification reaction was studied. The optimal conditions found for the kinetic resolution of m- and p-aryltrimethylsilyl chiral alcohols, led to excellent results, high conversions (c = 50%), high enantiomeric ratios (E > 200) and enantiomeric excesses for the remaining (S)-alcohol and (R)-acetylated product (>99%). However, kinetic resolution of o-aryltrimethylsilyl chiral alcohols did not occur under the same conditions applied to the other isomers.
Product Description
Supelco
Celite® 545 AW, reagent grade
Supelco
Celatom®, acid-washed, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Sigma-Aldrich
Filter agent, Celite® 545
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Diatomaceous earth