MilliporeSigma
  • Visual detection of organophosphorus pesticides represented by mathamidophos using Au nanoparticles as colorimetric probe.
Visual detection of organophosphorus pesticides represented by mathamidophos using Au nanoparticles as colorimetric probe.
Talanta (2011-11-22)
Hongkun Li, Jiajia Guo, Hong Ping, Lurui Liu, Minwei Zhang, Fengrui Guan, Chunyan Sun, Qian Zhang
ABSTRACT
With citrate-coated Au nanoparticles as colorimetric probe, a novel visual method for rapid assay of organophosphorus pesticides has been developed. The assay principle is based on catalytic hydrolysis of acetylthiocholine into thiocholine by acetylcholinesterase, which induces the aggregation of Au nanoparticles and the color change from claret-red to purple or even grey. The original plasmon absorption of Au nanoparticles at 522 nm decreases, and simultaneously, a new absorption band appears at 675 nm. The irreversible inhibition of organophosphorus pesticides on acetylcholinesterase prevents aggregation of Au nanoparticles. Under optimum conditions, the absorbance at 522 nm of Au nanoparticles is related linearly to the concentration of mathamidophos in the range of 0.02-1.42 μg/mL with a detection limit of 1.40 ng/mL. This colorimetric method has been successfully utilized to detect mathamidophos in vegetables with satisfactory results. The proposed colorimetric assay exhibits good reproducibility and accuracy, providing a simple and rapid method for the analysis of organophosphorus pesticides.
Product Description
Supelco
Methamidophos, PESTANAL®, analytical standard
Pricing and availability is not currently available.