MilliporeSigma
  • Fracture strength of incisor crowns after intracoronal bleaching with sodium percarbonate.
Fracture strength of incisor crowns after intracoronal bleaching with sodium percarbonate.
Dental traumatology : official publication of International Association for Dental Traumatology (2011-11-22)
Milton Carlos Kuga, José Mauricio dos Santos Nunes Reis, Semíramis Fabrício, Idomeo Bonetti-Filho, Edson Alves de Campos, Gisele Faria
ABSTRACT
To compare the fracture resistance of bovine teeth after intracoronal bleaching with sodium percarbonate (SPC) or sodium perborate (SP) mixed with water or 20% hydrogen peroxide (HP). Fifty extracted bovine teeth were divided into four experimental groups (G1-G4) and one control (n = 10) after endodontic treatment. Following root canal obturation, a glass ionomer barrier was placed at the cemento-enamel junction. After that, the pulp chambers were filled with: G1 - SP with water; G2 - SP with 20% HP; G3 - SPC with water; and G4 - SPC with 20% HP. No bleaching agent was used in the control group. Coronal access cavities were sealed with glass ionomer and specimens were immersed in artificial saliva. The bleaching agents were replaced after 7 days, and teeth were kept in artificial saliva for an additional 7 days, after which the pastes were removed and the coronal access cavities were restored with glass ionomer. Crowns were subjected to compressive load at a cross head speed of 0.5 mm min(-1) applied at 135° to the long axis of the root by an EMIC DL2000 testing machine, until coronal fracture. Data were statistically analysed by anova and Tukey test. No differences in fracture resistance were observed between the experimental groups (P > 0.05). However, all experimental groups presented lower fracture resistance than the control group (P < 0.05). SPC and SP led to equal reduction on fracture resistance of dental crowns, regardless of being mixed with water or 20% HP.
Sigma-Aldrich
Sodium perborate monohydrate, powder, 20-100 mesh
Sigma-Aldrich
Sodium perborate tetrahydrate, 9-11% available Oxygen
Sigma-Aldrich
Sodium perborate tetrahydrate, purum p.a., 96% (RT)