MilliporeSigma
  • Identification of 1,3-diiminoisoindoline carbohydrazides as potential antimalarial candidates.
Identification of 1,3-diiminoisoindoline carbohydrazides as potential antimalarial candidates.
ChemMedChem (2011-11-19)
Paolo Mombelli, Matthias C Witschel, Anthoni W van Zijl, Julie G Geist, Matthias Rottmann, Céline Freymond, Franz Röhl, Marcel Kaiser, Victoria Illarionova, Markus Fischer, Isabella Siepe, W Bernd Schweizer, Reto Brun, François Diederich
ABSTRACT
A series of inhibitors of plant enzymes of the non-mevalonate pathway from herbicide research efforts at BASF were screened for antimalarial activity in a cell-based assay. A 1,3-diiminoisoindoline carbohydrazide was found to inhibit the growth of Plasmodium falciparum with an IC(50) value <100 nM. Synthesis of a variety of derivatives allowed an improvement of the initial antimalarial activity down to IC(50) =18 nM for the most potent compound, the establishment of a structure-activity relationship, and the evaluation of the cytotoxic profile of the diiminoisoindolines. Furthermore, interesting configurational and conformational aspects for this class of compounds were studied by computational and X-ray crystal structure analysis. Some of the compounds can act as tridentate ligands, forming 2:1 ligand-iron(III) complexes, which also display antimalarial activity in the nanomolar IC(50) range, paired with low cytotoxicity.
Sigma-Aldrich
Carbohydrazide, 98%