MilliporeSigma
  • Efficient synthesis of biazoles by aerobic oxidative homocoupling of azoles catalyzed by a copper(I)/2-pyridonate catalytic system.
Efficient synthesis of biazoles by aerobic oxidative homocoupling of azoles catalyzed by a copper(I)/2-pyridonate catalytic system.
Chemical communications (Cambridge, England) (2011-11-15)
Mingwen Zhu, Ken-ichi Fujita, Ryohei Yamaguchi
ABSTRACT
A highly efficient and convenient CuCl/2-pyridonate catalytic system for oxidative homocoupling of azoles affording a biazole product has been developed. With this system, a variety of biazoles have been effectively synthesized in good to excellent yields in the presence of a very small amount of copper catalyst (1.0 mol%). It was feasible to employ air as a green oxidant.
Product Description
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Hydroxypyridine, 97%